Assay Details
| Gene Symbol: LOC4338083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783799.2 | XP_015639285.1 | 1 | 1796 | 148 | |
| GenBank mRNA | AK107068.1 | - | 1 | 1748 | 148 | |
More Information
Set Membership: |
|

LOC4338083
Genomic Map