Assay Details
| Gene Symbol: LOC4339725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782637.2 | XP_015638123.1 | 1 | 2136 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4339725| Gene Symbol: LOC4339725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782637.2 | XP_015638123.1 | 1 | 2136 | 66 | |
Set Membership: |
|