Assay Details
| Gene Symbol: LOC4344297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791381.2 | XP_015646867.1 | 17 - 18 | 4839 | 92 | |
| XM_015791382.2 | XP_015646868.1 | 17 - 18 | 4782 | 92 | ||
| GenBank mRNA | AK070164.1 | - | 1 - 2 | 349 | 92 | |
More Information
Set Membership: |
|

LOC4344297
Genomic Map