Assay Details
| Gene Symbol: LOC4345459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795350.2 | XP_015650836.1 | 2 - 3 | 510 | 74 | |
| XM_015795352.2 | XP_015650838.1 | 2 - 3 | 532 | 74 | ||
| GenBank mRNA | AK073889.1 | - | 2 - 3 | 502 | 74 | |
| AK119576.1 | - | 2 - 3 | 537 | 74 | ||
More Information
Set Membership: |
|

LOC4345459
Genomic Map