Assay Details
| Gene Symbol: LOC4345459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795350.2 | XP_015650836.1 | 7 - 8 | 1058 | 64 | |
| XM_015795352.2 | XP_015650838.1 | 7 - 8 | 1080 | 64 | ||
| GenBank mRNA | AK073889.1 | - | 7 - 8 | 1050 | 64 | |
| AK119576.1 | - | 7 - 8 | 1085 | 64 | ||
More Information
Set Membership: |
|

LOC4345459
Genomic Map