Assay Details
| Gene Symbol: LOC4345459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795350.2 | XP_015650836.1 | 8 - 9 | 1205 | 75 | |
| XM_015795352.2 | XP_015650838.1 | 8 - 9 | 1227 | 75 | ||
| GenBank mRNA | AK073889.1 | - | 8 - 9 | 1197 | 75 | |
| AK119576.1 | - | 8 - 9 | 1232 | 75 | ||
More Information
Set Membership: |
|

LOC4345459
Genomic Map