Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 11 - 12 | 1112 | 91 | |
| GenBank mRNA | AK099980.1 | - | 8 - 9 | 735 | 91 | |
| AK102066.1 | - | 11 - 12 | 1043 | 91 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map