Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 12 - 13 | 1270 | 98 | |
| GenBank mRNA | AK099980.1 | - | 9 - 10 | 893 | 98 | |
| AK102066.1 | - | 12 - 13 | 1201 | 98 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map