Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 13 - 14 | 1330 | 79 | |
| GenBank mRNA | AK099980.1 | - | 10 - 11 | 953 | 79 | |
| AK102066.1 | - | 13 - 14 | 1261 | 79 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map