Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 18 - 19 | 1891 | 76 | |
| XM_015791253.2 | XP_015646739.1 | 16 - 17 | 1681 | 76 | ||
| GenBank mRNA | AK102066.1 | - | 18 - 19 | 1822 | 76 | |
More Information
Set Membership: |
|

LOC4342584
Genomic Map