Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 4 - 5 | 490 | 131 | |
| XM_015791253.2 | XP_015646739.1 | 4 - 5 | 490 | 131 | ||
| GenBank mRNA | AK099980.1 | - | 1 - 2 | 96 | 131 | |
| AK102066.1 | - | 4 - 5 | 421 | 131 | ||
| AK102995.1 | - | 4 - 5 | 418 | 131 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map