Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 5 - 6 | 611 | 102 | |
| XM_015791253.2 | XP_015646739.1 | 5 - 6 | 611 | 102 | ||
| GenBank mRNA | AK099980.1 | - | 2 - 3 | 217 | 102 | |
| AK102066.1 | - | 5 - 6 | 542 | 102 | ||
| AK102995.1 | - | 5 - 6 | 539 | 102 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map