Assay Details
| Gene Symbol: LOC4342584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791252.2 | XP_015646738.1 | 9 - 10 | 997 | 144 | |
| XM_015791253.2 | XP_015646739.1 | 9 - 10 | 997 | 144 | ||
| GenBank mRNA | AK099980.1 | - | 6 - 7 | 620 | 144 | |
| AK102066.1 | - | 9 - 10 | 928 | 144 | ||
| AK102995.1 | - | 9 - 10 | 925 | 144 | ||
More Information
Set Membership: |
|

LOC4342584
Genomic Map