Assay Details
Gene Symbol: LOC4345577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794931.2 | XP_015650417.1 | 1 - 2 | 593 | 74 | |
GenBank mRNA | AK070406.1 | - | 1 - 2 | 599 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
