Assay Details
Gene Symbol: LOC4345577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794931.2 | XP_015650417.1 | 6 - 7 | 2017 | 88 | |
GenBank mRNA | AK070406.1 | - | 6 - 7 | 2023 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
