Assay Details
| Gene Symbol: LOC4345577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794931.2 | XP_015650417.1 | 9 - 10 | 2206 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4345577| Gene Symbol: LOC4345577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794931.2 | XP_015650417.1 | 9 - 10 | 2206 | 150 | |
Set Membership: |
|