Assay Details
| Gene Symbol: LOC4345625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795563.2 | XP_015651049.1 | 4 - 5 | 840 | 78 | |
| XM_015795564.2 | XP_015651050.1 | 4 - 5 | 825 | 78 | ||
| GenBank mRNA | AK103459.1 | - | 4 - 5 | 801 | 78 | |
| HQ858803.1 | - | 4 - 5 | 662 | 78 | ||
More Information
Set Membership: |
|

LOC4345625
Genomic Map