Assay Details
| Gene Symbol: LOC4346090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794631.2 | XP_015650117.1 | 10 - 11 | 1601 | 92 | |
| GenBank mRNA | AK103172.1 | - | 10 - 11 | 1543 | 92 | |
More Information
Set Membership: |
|

LOC4346090
Genomic Map