Assay Details
Gene Symbol: LOC4346747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795965.2 | XP_015651451.1 | 2 - 3 | 284 | 72 | |
GenBank mRNA | AK068681.1 | - | 2 - 3 | 239 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
