Assay Details
| Gene Symbol: LOC4346747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795965.2 | XP_015651451.1 | 5 - 6 | 573 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4346747| Gene Symbol: LOC4346747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795965.2 | XP_015651451.1 | 5 - 6 | 573 | 73 | |
Set Membership: |
|