Assay Details
| Gene Symbol: LOC4347617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755130.2 | XP_015610616.1 | 1 - 2 | 336 | 151 | |
| GenBank mRNA | AB158320.1 | - | 1 - 2 | 302 | 151 | |
| AK063534.1 | - | 1 - 2 | 330 | 151 | ||
More Information
Set Membership: |
|

LOC4347617
Genomic Map