Assay Details
| Gene Symbol: LOC4349076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759028.2 | XP_015614514.1 | 16 - 17 | 3639 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4349076| Gene Symbol: LOC4349076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759028.2 | XP_015614514.1 | 16 - 17 | 3639 | 68 | |
Set Membership: |
|