Assay Details
| Gene Symbol: LOC4349924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761516.2 | XP_015617002.1 | 11 - 12 | 2576 | 118 | |
| GenBank mRNA | AK108748.1 | - | 10 - 11 | 1422 | 118 | |
| AK121851.1 | - | 11 - 12 | 2584 | 118 | ||
| AK122056.1 | - | 11 - 12 | 2574 | 118 | ||
More Information
Set Membership: |
|

LOC4349924
Genomic Map