Assay Details
| Gene Symbol: LOC4349924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761516.2 | XP_015617002.1 | 12 - 13 | 2738 | 83 | |
| GenBank mRNA | AK108748.1 | - | 11 - 12 | 1584 | 83 | |
| AK121851.1 | - | 12 - 13 | 2746 | 83 | ||
| AK122056.1 | - | 12 - 13 | 2736 | 83 | ||
More Information
Set Membership: |
|

LOC4349924
Genomic Map