Assay Details
| Gene Symbol: LOC4349924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761516.2 | XP_015617002.1 | 14 - 15 | 2979 | 101 | |
| GenBank mRNA | AK121851.1 | - | 14 - 15 | 2987 | 101 | |
| AK122056.1 | - | 14 - 15 | 2977 | 101 | ||
More Information
Set Membership: |
|

LOC4349924
Genomic Map