Assay Details
| Gene Symbol: LOC4349924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761516.2 | XP_015617002.1 | 3 - 4 | 1581 | 73 | |
| GenBank mRNA | AK108748.1 | - | 2 - 3 | 427 | 73 | |
| AK121851.1 | - | 3 - 4 | 1589 | 73 | ||
| AK122056.1 | - | 3 - 4 | 1579 | 73 | ||
More Information
Set Membership: |
|

LOC4349924
Genomic Map