Assay Details
| Gene Symbol: LOC4352549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765112.2 | XP_015620598.1 | 10 - 11 | 1331 | 75 | |
| GenBank mRNA | AK062236.1 | - | 10 - 11 | 1337 | 75 | |
| AK071114.1 | - | 10 - 11 | 1338 | 75 | ||
More Information
Set Membership: |
|

LOC4352549
Genomic Map