Assay Details
| Gene Symbol: LOC4352549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765112.2 | XP_015620598.1 | 2 - 3 | 404 | 98 | |
| GenBank mRNA | AK062236.1 | - | 2 - 3 | 410 | 98 | |
| AK071114.1 | - | 2 - 3 | 411 | 98 | ||
More Information
Set Membership: |
|

LOC4352549
Genomic Map