Assay Details
| Gene Symbol: LOC4352549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765112.2 | XP_015620598.1 | 7 - 8 | 999 | 83 | |
| GenBank mRNA | AK062236.1 | - | 7 - 8 | 1005 | 83 | |
| AK071114.1 | - | 7 - 8 | 1006 | 83 | ||
More Information
Set Membership: |
|

LOC4352549
Genomic Map