Assay Details
| Gene Symbol: LOC4352684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764836.2 | XP_015620322.1 | 6 - 7 | 2065 | 101 | |
| GenBank mRNA | AK060570.1 | - | 1 - 2 | 227 | 101 | |
| AK067863.1 | - | 7 - 8 | 1439 | 101 | ||
More Information
Set Membership: |
|

LOC4352684
Genomic Map