Assay Details
| Gene Symbol: LOC4352684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764836.2 | XP_015620322.1 | 7 - 8 | 2225 | 79 | |
| GenBank mRNA | AK060570.1 | - | 2 - 3 | 387 | 79 | |
| AK067863.1 | - | 8 - 9 | 1599 | 79 | ||
More Information
Set Membership: |
|

LOC4352684
Genomic Map