Assay Details
| Gene Symbol: LOC4350026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761693.2 | XP_015617179.1 | 1 | 1539 | 126 | |
| GenBank mRNA | AK100873.1 | - | 1 | 1441 | 126 | |
More Information
Set Membership: |
|

LOC4350026
Genomic Map