Assay Details
| Gene Symbol: LOC4330049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770913.1 | XP_015626399.1 | 4 | 1479 | 144 | |
| GenBank mRNA | AK061782.1 | - | 4 | 1147 | 144 | |
| AK120386.1 | - | 2 | 507 | 144 | ||
| BT131322.1 | - | 263 | 144 | |||
More Information
Set Membership: |
|

LOC4330049
Genomic Map