Assay Details
| Gene Symbol: LOC4330832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767283.2 | XP_015622769.1 | 4 | 1462 | 162 | |
| GenBank mRNA | AK101744.1 | - | 4 | 1448 | 162 | |
| EU267974.1 | - | 4 | 1435 | 162 | ||
More Information
Set Membership: |
|

LOC4330832
Genomic Map