Assay Details
| Gene Symbol: LOC4333619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776195.1 | XP_015631681.1 | 9 | 1045 | 117 | |
| XM_015776196.2 | XP_015631682.1 | 9 | 1174 | 117 | ||
| GenBank mRNA | AK061103.1 | - | 9 | 955 | 117 | |
| AK099162.1 | - | 9 | 946 | 117 | ||
More Information
Set Membership: |
|

LOC4333619
Genomic Map