Assay Details
| Gene Symbol: LOC4341623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787244.2 | XP_015642730.1 | 2 | 1651 | 98 | |
| GenBank mRNA | AK060486.1 | - | 2 | 1651 | 98 | |
| AK106068.1 | - | 2 | 1637 | 98 | ||
More Information
Set Membership: |
|

LOC4341623
Genomic Map