Assay Details
| Gene Symbol: LOC9272332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761803.2 | XP_015617289.1 | 8 | 3532 | 86 | |
| XM_015761804.1 | XP_015617290.1 | 8 | 1605 | 86 | ||
| XM_015761805.1 | XP_015617291.1 | 8 | 1595 | 86 | ||
| Gene Symbol: LOC4351241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764296.1 | XP_015619782.1 | 8 | 1605 | 86 | |
| XM_015764297.1 | XP_015619783.1 | 8 | 1599 | 86 | ||
| XM_015764298.1 | XP_015619784.1 | 9 | 1785 | 86 | ||
| GenBank mRNA | AK068187.1 | - | 6 | 1352 | 86 | |
More Information
Set Membership: |
|

LOC9272332
Genomic Map