Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261057.1 | NP_001247986.1 | 842 | 116 | ||
| XM_015147638.2 | XP_015003124.1 | 713 | 116 | |||
| XM_015147639.2 | XP_015003125.1 | 857 | 116 | |||
| GenBank mRNA | JU320743.1 | - | 842 | 116 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LGI1
Genomic Map