Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204121.2 | NP_001191050.1 | 449 | 84 | ||
| NM_001371502.1 | NP_001358431.1 | 323 | 84 | |||
| GenBank mRNA | JU332794.1 | - | 323 | 84 | ||
| JU334592.1 | - | 323 | 84 | |||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map