Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194684.2 | NP_001181613.1 | 590 | 84 | ||
| XM_015152033.2 | XP_015007519.1 | 608 | 84 | |||
| XM_015152034.2 | XP_015007520.1 | 744 | 84 | |||
| XM_028828983.1 | XP_028684816.1 | 652 | 84 | |||
| XM_028828984.1 | XP_028684817.1 | 907 | 84 | |||
| XM_028828985.1 | XP_028684818.1 | 901 | 84 | |||
| GenBank mRNA | JU336878.1 | - | 691 | 84 | ||
| JU477617.1 | - | 689 | 84 | |||
| JV485493.1 | - | 509 | 84 | |||
| JV596976.1 | - | 505 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSD17B6
Genomic Map