Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114101.4 | XP_001114101.3 | 390 | 86 | ||
| XM_015124658.2 | XP_014980144.2 | 387 | 86 | |||
| XM_015124659.2 | XP_014980145.2 | 257 | 86 | |||
| XM_015124660.2 | XP_014980146.2 | 287 | 86 | |||
| XM_028839819.1 | XP_028695652.1 | 315 | 86 | |||
| GenBank mRNA | JU332258.1 | - | 162 | 86 | ||
| JU473143.1 | - | 164 | 86 | |||
More Information
Set Membership: |
|

JOSD2
Genomic Map