Assay Details
| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265908.1 | NP_001252837.1 | 844 | 83 | ||
| GenBank mRNA | JV044556.1 | - | 803 | 83 | ||
| JV635689.1 | - | 844 | 83 | |||
More Information
Set Membership: |
|

GNA14
Genomic Map