Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088894.4 | XP_001088894.2 | 253 | 77 | ||
XM_002803820.3 | XP_002803866.1 | 249 | 77 | |||
XM_028847376.1 | XP_028703209.1 | 498 | 77 | |||
GenBank mRNA | JU471998.1 | - | 178 | 77 | ||
JV045211.1 | - | 198 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
