Assay Details
| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260669.1 | NP_001247598.1 | 768 | 67 | ||
| XM_015143349.2 | XP_014998835.2 | 1247 | 67 | |||
| GenBank mRNA | JU332486.1 | - | 763 | 67 | ||
| JU473958.1 | - | 768 | 67 | |||
| JV640105.1 | - | 777 | 67 | |||
More Information
Set Membership: |
|

IRF9
Genomic Map