Assay Details
| Gene Symbol: CASQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148912.2 | XP_015004398.2 | 699 | 96 | ||
| XM_015148921.2 | XP_015004407.2 | 597 | 96 | |||
| XM_028830069.1 | XP_028685902.1 | 699 | 96 | |||
More Information
Set Membership: |
|

CASQ2
Genomic Map