Assay Details
| Gene Symbol: IL15RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079517.1 | NP_001072985.1 | 481 | 83 | ||
| NM_001172844.1 | NP_001166315.1 | 580 | 83 | |||
| XM_015146368.2 | XP_015001854.2 | 678 | 83 | |||
| XM_015146369.2 | XP_015001855.2 | 579 | 83 | |||
| XM_015146370.2 | XP_015001856.1 | 480 | 83 | |||
| XM_015146371.2 | XP_015001857.1 | 903 | 83 | |||
| XM_028853491.1 | XP_028709324.1 | 804 | 83 | |||
| GenBank mRNA | EF033114.1 | - | 580 | 83 | ||
| EF033115.1 | - | 481 | 83 | |||
| EF033116.1 | - | 382 | 83 | |||
| FJ222743.1 | - | 499 | 83 | |||
| FJ222744.1 | - | 301 | 83 | |||
| JU485931.1 | - | 359 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL15RA
Genomic Map