Assay Details
| Gene Symbol: EPHX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098374.4 | XP_001098374.1 | 438 | 99 | ||
| XM_028831409.1 | XP_028687242.1 | 206 | 99 | |||
| GenBank mRNA | JV463266.1 | - | 156 | 99 | ||
| JV611756.1 | - | 423 | 99 | |||
More Information
Set Membership: |
|

EPHX4
Genomic Map