Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193876.2 | NP_001180805.1 | 1771 | 78 | ||
| XM_001090655.4 | XP_001090655.1 | 2033 | 78 | |||
| XM_015117346.2 | XP_014972832.1 | 2104 | 78 | |||
| XM_015117348.2 | XP_014972834.1 | 1813 | 78 | |||
| XM_015117349.2 | XP_014972835.1 | 1950 | 78 | |||
| XM_015117350.2 | XP_014972836.1 | 1741 | 78 | |||
| XM_028834236.1 | XP_028690069.1 | 1751 | 78 | |||
| XM_028834237.1 | XP_028690070.1 | 2329 | 78 | |||
| GenBank mRNA | JU321302.1 | - | 1662 | 78 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FAM214B
Genomic Map