Assay Details
| Gene Symbol: TGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799271.3 | XP_002799317.1 | 600 | 70 | ||
| XM_015112609.2 | XP_014968095.1 | 608 | 70 | |||
| GenBank mRNA | JU328393.1 | - | 247 | 70 | ||
| S75913.1 | - | 300 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TGFA
Genomic Map