Assay Details
| Gene Symbol: IRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267018.1 | NP_001253947.1 | 636 | 97 | ||
| XM_015116861.2 | XP_014972347.1 | 553 | 97 | |||
| GenBank mRNA | JU472495.1 | - | 636 | 97 | ||
| JV060209.1 | - | 644 | 97 | |||
More Information
Set Membership: |
|

IRF6
Genomic Map